missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Tiglate 98.0+%, TCI America™

Catalog No. T024725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T024725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T024725G Supplier TCI America Supplier No. T024725G

Chemical Identifiers

CAS 5837-78-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015183
InChI Key OAPHLAAOJMTMLY-GQCTYLIASA-N
Synonym ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate
PubChem CID 5281163
ChEBI CHEBI:4892
IUPAC Name ethyl (E)-2-methylbut-2-enoate
SMILES CCOC(=O)C(=CC)C

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 g
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Tiglate
EINECSNumber (2)-0969
TSCA Yes