missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tiglate, 98%

Catalog No. AAA1202922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1202922 100 g
AAA1202914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1202922 Supplier Thermo Scientific Chemicals Supplier No. A1202922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5837-78-5 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5837-78-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015183
InChI Key OAPHLAAOJMTMLY-GQCTYLIASA-N
Synonym ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate
PubChem CID 5281163
ChEBI CHEBI:4892
IUPAC Name ethyl (E)-2-methylbut-2-enoate
SMILES CCOC(=O)C(=CC)C

Specifications

Density 0.93
Boiling Point 156°C
Flash Point 46°C (114°F)
Refractive Index 1.434
Linear Formula CH3CH=C(CH3)CO2CH2CH3
Quantity 100 g
UN Number UN3272
Beilstein 1720895
Merck Index 14,9433
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Ethyl tiglate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 227-425-6
RTECSNumber EM9252700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only