missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tetradecanoate, 98%

Catalog No. AAA11646AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11646AP 500 mL
AAA11646AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11646AP Supplier Thermo Scientific Chemicals Supplier No. A11646AP
Only null left

CAS: 124-06-1 | C16H32O2 | 256.43 g/mol

A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 124-06-1
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00008984
InChI Key MMKRHZKQPFCLLS-UHFFFAOYSA-N
Synonym ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
PubChem CID 31283
ChEBI CHEBI:84849
IUPAC Name ethyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC

Specifications

Melting Point 12°C
Density 0.86
Boiling Point 295°C
Flash Point 135°C (275°F)
Odor Weak
Refractive Index 1.436
Linear Formula CH3(CH2)12CO2CH2CH3
Quantity 500 mL
Beilstein 1776382
Merck Index 14,6333
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 256.43
Percent Purity 98%
Chemical Name or Material Ethyl tetradecanoate
Show More Show Less
EINECSNumber 204-675-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only