missing translation for 'onlineSavingsMsg'
Learn More

Ethyl sorbate, 96%

Catalog No. AAA1143136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1143136 500 g
AAA1143114 25 g
AAA1143122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1143136 Supplier Thermo Scientific Chemicals Supplier No. A1143136

CAS: 2396-84-1 | C8H12O2 | 140.18 g/mol

The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.

Solubility
Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2396-84-1
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00009296
InChI Key OZZYKXXGCOLLLO-TWTPFVCWSA-N
Synonym ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459
PubChem CID 1550470
IUPAC Name ethyl (2E,4E)-hexa-2,4-dienoate
SMILES CCOC(=O)\C=C\C=C\C

Specifications

Density 0.948
Boiling Point 195°C to 196°C
Flash Point 69°C (156°F)
Refractive Index 1.495
Quantity 500 g
Beilstein 1362941
Solubility Information Soluble in alcohol.
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material Ethyl sorbate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 219-258-2
RTECSNumber WG2143000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only