missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-4-Chloro-3-hydroxybutyrate 95.0+%, TCI America™

Catalog No. C17175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17175G 5 g
C171725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17175G Supplier TCI America Supplier No. C17175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86728-85-0
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.60
MDL Number MFCD00211241
InChI Key ZAJNMXDBJKCCAT-YFKPBYRVSA-N
Synonym ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate
PubChem CID 7019277
IUPAC Name ethyl (3S)-4-chloro-3-hydroxybutanoate
SMILES CCOC(=O)C[C@H](O)CCl

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 166.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (S)-4-Chloro-3-hydroxybutyrate
EINECSNumber (2)-3782&(2)-3796&(2)-3882
TSCA No
Recommended Storage Refrigerator