missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthyle (S)-(+)-3-pipéridinecarboxylate 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Quantity | 5 g |
| Formula Weight | 157.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | Ethyl (S)-(+)-3-Piperidinecarboxylate |
Sécurité et manipulation
| TSCA | No |