missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-(-)-3-Hydroxybutyrate 98.0+%, TCI America™

Catalog No. H10295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10295G 5 g
H10291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10295G Supplier TCI America Supplier No. H10295G

Chemical Identifiers

CAS 24915-95-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00075386
InChI Key OMSUIQOIVADKIM-RXMQYKEDSA-N
Synonym ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester
PubChem CID 440030
ChEBI CHEBI:28707
IUPAC Name ethyl (3R)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-(-)-3-Hydroxybutyrate
EINECSNumber (2)-1430
TSCA No