missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 24915-95-5 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00075386 |
| InChI Key | OMSUIQOIVADKIM-RXMQYKEDSA-N |
| Synonym | ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester |
| PubChem CID | 440030 |
| ChEBI | CHEBI:28707 |
| IUPAC Name | ethyl (3R)-3-hydroxybutanoate |
| SMILES | CCOC(=O)CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC345760010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC345760050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 24915-95-5 | |
| 132.16 | |
| OMSUIQOIVADKIM-RXMQYKEDSA-N | |
| 440030 | |
| ethyl (3R)-3-hydroxybutanoate |
| C6H12O3 | |
| MFCD00075386 | |
| ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester | |
| CHEBI:28707 | |
| CCOC(=O)CC(C)O |
Specifications
| 24915-95-5 | |
| 75°C to 76°C (12.0mmHg) | |
| 97.5% min. (GC) | |
| CH3CH(OH)CH2COOCH2CH3 | |
| MFCD00075386 | |
| −45.5° (20°C 589nm,c=1,CHCl3) | |
| ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester | |
| OMSUIQOIVADKIM-RXMQYKEDSA-N | |
| ethyl (3R)-3-hydroxybutanoate | |
| 440030 | |
| 132.16 | |
| Ethyl (R)-3-hydroxybutyrate, 98% |
| 1.0170g/mL | |
| Authentic | |
| C6H12O3 | |
| 1.4195 to 1.4215 | |
| 1.017 | |
| -45.5 | |
| Solubility in water: soluble in water | |
| CCOC(=O)CC(C)O | |
| 132.16 | |
| CHEBI:28707 | |
| 98% |
RUO – Research Use Only