missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propenyl ether, cis + trans, 97%

Catalog No. AAH2631922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2631922 100 g
AAH2631906 5 g
AAH2631914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2631922 Supplier Thermo Scientific Chemicals Supplier No. H2631922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 928-55-2 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-55-2
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00015184
InChI Key XDHOEHJVXXTEDV-HYXAFXHYSA-N
Synonym 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
PubChem CID 5365091
SMILES CCO\C=C/C

Specifications

Density 0.778
Boiling Point 69°C to 71°C
Flash Point 20°C (68°F)
Refractive Index 1.398
Quantity 100 g
UN Number UN3271
Beilstein 1735974
IUPAC Name (E)-1-ethoxyprop-1-ene
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material Ethyl propenyl ether, cis + trans
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 213-176-0
RTECSNumber KO0525000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only