missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phenylpropiolate 97.0+%, TCI America™

Catalog No. P08145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08145G 5 g
P081425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08145G Supplier TCI America Supplier No. P08145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2216-94-6
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00009185
InChI Key ACJOYTKWHPEIHW-UHFFFAOYSA-N
Synonym ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate
PubChem CID 91516
IUPAC Name ethyl 3-phenylprop-2-ynoate
SMILES CCOC(=O)C#CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
Formula Weight 174.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Phenylpropiolate
RTECSNumber UE0110000
TSCA No
Recommended Storage Refrigerator