missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthyle p-toluènesulfonate 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Blanc-jaune |
| Boiling Point | 173°C |
| Quantity | 500 g |
| Formula Weight | 200.25 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Morceaux cristallins |
| Chemical Name or Material | Ethyl p-Toluenesulfonate |
Sécurité et manipulation
| EINECSNumber | (3)-1896 |
| RTECSNumber | XT6825000 |
| TSCA | Yes |