missing translation for 'onlineSavingsMsg'
Learn More

Ethyl myristate, 98+%, Thermo Scientific™

Numéro de catalogue. p-3586429
Modifier l'affichage
Click to view available options
Quantité:
25g
100g
500g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC385540250 25g Glass bottle
AC385541000 100g Glass bottle
AC385545000 500g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC385540250 Fournisseur Thermo Scientific Chemicals Code fournisseur 385540250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 124-06-1
Formule moléculaire C16H32O2
Poids moléculaire (g/mol) 256.43
Numéro MDL MFCD00008984
Clé InChI MMKRHZKQPFCLLS-UHFFFAOYSA-N
Synonyme ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
PubChem CID 31283
ChEBI CHEBI:84849
Nom de l’IUPAC ethyl tetradecanoate
SOURIRES CCCCCCCCCCCCCC(=O)OCC

Spécifications

Nom ou substance chimique Ethyl myristate
Densité 0.8610g/mL
Point d’ébullition 295.0°C
Point d’éclair >110°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98% min. (GC)
Emballage Glass bottle
Formule linéaire CH3(CH2)12COOCH2CH3
Quantité 25g
Gravité spécifique 0.861
Indice de Merck 15, 6419
Renseignements sur la solubilité Solubility in water: insoluble
Poids de formule 256.43
Pourcentage de pureté 98+%
Afficher plus de résultats Afficher moins de résultats
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-675-4

RUO – Research Use Only