missing translation for 'onlineSavingsMsg'
Learn More

Ethyl maltol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014311
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014311 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014311 Supplier Merck Emd Millipore Supplier No. 94185100MG

Ethyl maltol is a synthetic homologue of maltol, often found as flavor enhancers and it contributes to the fragrance of commercials products such as cereals, breads, malt beverages, coffee, soybeans and chocolate milk.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 11-8-4940
Physical Form Neat
Linear Formula C7H8O3
MDL Number MFCD00059795
Melting Point 85°C to 95°C
Synonym 2-Ethyl-3-hydroxy-4 H-pyran-4-one; Ethyl maltol
Type Analytical Standard
Quantity 100 mg
Formula Weight 140.14
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less