missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Lactate 98.0+%, TCI America™

Catalog No. L000325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L000325ML 25 mL
L0003500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L000325ML Supplier TCI America Supplier No. L000325ML
Only null left

Chemical Identifiers

CAS 97-64-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00065359
InChI Key LZCLXQDLBQLTDK-UHFFFAOYSA-N
Synonym ethyl lactate, actylol, acytol, solactol, lactic acid, ethyl ester, propanoic acid, 2-hydroxy-, ethyl ester, lactate d'ethyle, 2-hydroxypropanoic acid ethyl ester, ethyl 2-hydroxypropionate, ethyl alpha-hydroxypropionate
PubChem CID 7344
ChEBI CHEBI:78321
IUPAC Name ethyl 2-hydroxypropanoate
SMILES CCOC(=O)C(C)O

Specifications

Melting Point -25°C
Color Colorless
Boiling Point 154°C
Quantity 25 mL
UN Number 1192
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Lactate
EINECSNumber (2)-1371
RTECSNumber OD5075000
TSCA Yes