missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-Lactate, 98%, Spectrum™ Chemical

$186.91 - $21032.97

Chemical Identifiers

CAS 97-64-3
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610432
View Documents
Spectrum Chemical Mfg Cor
E1187100ML
100 mL Amber Glass Bottle
Each for $186.91
Only null left
 
18610436
View Documents
Spectrum Chemical Mfg Cor
E1187500MLGL
500 mL 98%
Each for $523.44
Only null left
 
18610435
View Documents
Spectrum Chemical Mfg Cor
E11874LTGL
4 L 98%
Each for $1,429.91
Only null left
 
18610433
View Documents
Spectrum Chemical Mfg Cor
E118720LT
20 L 98%
Each for $4,113.94
Only null left
 
18610434
View Documents
Spectrum Chemical Mfg Cor
E1187200LTBL
200 L 98% N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

97-64-3

Specifications

Amber Glass Bottle
1
98%
Ethyl L-Lactate
97-64-3
100 mL
Reagent
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.