CAS RN 97-64-3
CAS RN 97-64-3
IUPAC Name:
ethyl 2-hydroxypropanoate
Synonyms:
ethyl lactate
Molecular Weight (g/mol):
118.13
Molecular Formula:
C5H10O3
InChi Key:
LZCLXQDLBQLTDK-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(C)O
1
–
2
of
2
results
1
Ethyl Lactate 98.0+%, TCI America™
CAS: 97-64-3 Molecular Formula: C5H10O3 Molecular Weight (g/mol): 118.132 MDL Number: MFCD00065359 InChI Key: LZCLXQDLBQLTDK-UHFFFAOYSA-N Synonym: ethyl lactate,actylol,acytol,solactol,lactic acid, ethyl ester,propanoic acid, 2-hydroxy-, ethyl ester,lactate d'ethyle,2-hydroxypropanoic acid ethyl ester,ethyl 2-hydroxypropionate,ethyl alpha-hydroxypropionate PubChem CID: 7344 ChEBI: CHEBI:78321 IUPAC Name: ethyl 2-hydroxypropanoate SMILES: CCOC(=O)C(C)O
2
Ethyl L-Lactate, 98%, Spectrum™ Chemical
CAS: 97-64-3