missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-Lactate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor E11874LTGL

 View more versions of this product

Catalog No. 18610435


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

98%
1
4 L
Reagent
97-64-3
Amber Glass Bottle
98%

Chemical Identifiers

97-64-3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.