missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isopropyl Sulfone 97.0+%, TCI America™

Catalog No. E04755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04755G 5 g
E047525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04755G Supplier TCI America Supplier No. E04755G
Only null left

Chemical Identifiers

CAS 4853-75-2
Molecular Formula C5H12O2S
Molecular Weight (g/mol) 136.209
MDL Number MFCD00050418
InChI Key RDKKQZIFDSEMNU-UHFFFAOYSA-N
PubChem CID 21719919
IUPAC Name 2-ethylsulfonylpropane
SMILES CCS(=O)(=O)C(C)C

Specifications

Color Yellow
Boiling Point 91°C
Quantity 5 g
Formula Weight 136.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isopropyl Sulfone