missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Heptadecanoate 97.0+%, TCI America™

Catalog No. H05265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05265G 5 g
H052625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05265G Supplier TCI America Supplier No. H05265G

Chemical Identifiers

CAS 14010-23-2
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.511
MDL Number MFCD00026658
InChI Key KNXMUFRWYNVISA-UHFFFAOYSA-N
Synonym Ethyl Margarate, Heptadecanoic Acid Ethyl Ester, Margaric Acid Ethyl Ester
PubChem CID 26397
IUPAC Name ethyl heptadecanoate
SMILES CCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Color White
Boiling Point 200°C
Quantity 5 g
Formula Weight 298.51
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material Ethyl Heptadecanoate
EINECSNumber (2)-0798
TSCA No