missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Glutaryl Chloride 98.0+%, TCI America™

Catalog No. E04515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04515G 5 g
E045125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04515G Supplier TCI America Supplier No. E04515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5205-39-0
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.612
MDL Number MFCD00013659
InChI Key KKJAQUGGQMCNJY-UHFFFAOYSA-N
Synonym ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester
PubChem CID 78881
IUPAC Name ethyl 5-chloro-5-oxopentanoate
SMILES CCOC(=O)CCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 3265
Formula Weight 178.61
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl Glutaryl Chloride