missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glutaryl chloride, 97%

Catalog No. AAH59443AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59443AA 5 mL
AAH59443AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59443AA Supplier Thermo Scientific Chemicals Supplier No. H59443AA
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5205-39-0 | C7H11ClO3 | 178.612 g/mol

Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5205-39-0
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.612
MDL Number MFCD00013659
InChI Key KKJAQUGGQMCNJY-UHFFFAOYSA-N
Synonym ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester
PubChem CID 78881
IUPAC Name ethyl 5-chloro-5-oxopentanoate
SMILES CCOC(=O)CCCC(=O)Cl

Specifications

Density 1.139
Boiling Point 68°C to 70°C (0.07 mmHg)
Refractive Index 1.443
Linear Formula ClCO(CH2)3COOC2H5
Quantity 5 mL
UN Number UN3265
Beilstein 1099817
Solubility Information Soluble in water.
Formula Weight 178.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl glutaryl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 226-000-2
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only