missing translation for 'onlineSavingsMsg'
Learn More

Ethyl ethynyl ether, 50 wt% solution in hexanes

Catalog No. AC307390250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307390250 25 g Glass bottle
AC307390010 1 g Glass bottle
AC307390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC307390250 Supplier Thermo Scientific Chemicals Supplier No. 307390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 927-80-0 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 927-80-0 , 92112-69-1
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00009247
InChI Key WMYNMYVRWWCRPS-UHFFFAOYSA-N
Synonym ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
PubChem CID 61239
IUPAC Name ethynoxyethane
SMILES CCOC#C

Specifications

Color Orange to Brown
Density 0.7180g/mL
Boiling Point 55.0°C to 56.0°C
Flash Point −28°C
Packaging Glass bottle
Linear Formula C2H5OC≡CH
Quantity 25 g
Specific Gravity 0.718
Formula Weight 70.09
Physical Form Liquid
Chemical Name or Material Ethyl ethynyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.
Suspected of damaging fertility.
May be fatal if swallowed and enters airways.
May cause dam
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 213-164-5
RTECSNumber KN9900000

RUO – Research Use Only