missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ethoxyacetate 95.0+%, TCI America™

Numéro de catalogue. E12375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E12375G 5 g
E123725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E12375G Fournisseur TCI America Code fournisseur E12375G
Il en reste null

Chemical Identifiers

CAS 817-95-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00040492
InChI Key CKSRFHWWBKRUKA-UHFFFAOYSA-N
Synonym ethyl ethoxyacetate, ethoxyacetic acid ethyl ester, acetic acid, ethoxy-, ethyl ester, ethoxy-acetic acid ethyl ester, ethoxyacetic acid, ethyl ester, acetic acid, 2-ethoxy-, ethyl ester, diethyl glycolate, ethyl ethoxylacetate, acmc-20akxi
PubChem CID 69955
IUPAC Name ethyl 2-ethoxyacetate
SMILES CCOCC(=O)OCC

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 g
UN Number 3272
Formula Weight 132.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Ethoxyacetate