missing translation for 'onlineSavingsMsg'
Learn More

Ethyl diethoxyacetate, 97+%

Catalog No. AAL0039022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0039022 100 g
AAL0039014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0039022 Supplier Thermo Scientific Chemicals Supplier No. L0039022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6065-82-3 | C8H16O4 | 176.212 g/mol

Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.

Solubility
Soluble in Dichloromethane, Ethyl Acetate and Methanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 6065-82-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00009231
InChI Key XCLBIKIQSCTANZ-UHFFFAOYSA-N
Synonym ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal
PubChem CID 80169
IUPAC Name ethyl 2,2-diethoxyacetate
SMILES CCOC(C(=O)OCC)OCC

Specifications

Density 0.988
Boiling Point 198°C to 200°C
Flash Point 85°C (185°F)
Refractive Index 1.41
Quantity 100 g
Beilstein 878579
Solubility Information Soluble in Dichloromethane,Ethyl Acetate and Methanol.
Formula Weight 176.21
Percent Purity ≥97%
Chemical Name or Material Ethyl diethoxyacetate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 227-999-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only