missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dibromofluoroacetate, 97%

Catalog No. AAL1649114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1649114 25 g
AAL1649106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1649114 Supplier Thermo Scientific Chemicals Supplier No. L1649114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 565-53-7 | C4H5Br2FO2 | 263.89 g/mol

Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 565-53-7
Molecular Formula C4H5Br2FO2
Molecular Weight (g/mol) 263.89
MDL Number MFCD00042068
InChI Key COYRDMUQLSIAEX-UHFFFAOYSA-N
Synonym ethyl dibromofluoroacetate, dibromofluoroacetic acid ethyl ester, ethyldibromofluoroacetate, 2,2-dibromo-1-ethoxy-2-fluoroethan-1-one, acmc-1aqw2, ethyl 2,2-dibromo-2-fluoracetate, ethyl 2,2-dibromo-2-fluoro-acetate
PubChem CID 2737168
IUPAC Name ethyl 2,2-dibromo-2-fluoroacetate
SMILES CCOC(=O)C(F)(Br)Br

Specifications

Density 1.894
Boiling Point 82°C (33 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4635
Linear Formula Br2CFCO2CH2CH3
Quantity 25 g
UN Number UN2922
Beilstein 1762048
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 263.89
Percent Purity 97%
Chemical Name or Material Ethyl dibromofluoroacetate
Show More Show Less
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only