missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dibromoacetate, 96%

Catalog No. AAB2320914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2320914 25 g
AAB2320906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2320914 Supplier Thermo Scientific Chemicals Supplier No. B2320914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 617-33-4 | C4H6Br2O2 | 245.898 g/mol

Ethyl dibromoacetate can be used to produce diphenoxy-acetic acid ethyl ester. It is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromoacetate can be used to produce diphenoxy-acetic acid ethyl ester. It is used in organic synthesis.

Solubility
Soluble in hexane, ether, CH2Cl2, acetone, EtOAc, ethanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 617-33-4
Molecular Formula C4H6Br2O2
Molecular Weight (g/mol) 245.898
MDL Number MFCD00041718
InChI Key NIJGVVHCUXNSLL-UHFFFAOYSA-N
Synonym ethyl dibromoacetate, acetic acid, dibromo-, ethyl ester, dibromoacetic acid, ethyl ester, acetic acid, 2,2-dibromo-, ethyl ester, ethyldibromoacetate, ethyl dibromo acetate, acmc-20akxm, aceticacid,dibromo-,ethylester, dibromoacetic acid ethyl ester, dibromo-acetic acid ethyl ester
PubChem CID 69237
IUPAC Name ethyl 2,2-dibromoacetate
SMILES CCOC(=O)C(Br)Br

Specifications

Density 1.902
Boiling Point 76°C to 78°C (12 mmHg)
Refractive Index 1.499
Linear Formula Br2CHCO2CH2CH3
Quantity 25 g
Solubility Information Soluble in hexane,ether,CH2Cl2,acetone,EtOAc,ethanol.
Formula Weight 245.9
Percent Purity 96%
Chemical Name or Material Ethyl dibromoacetate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-510-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only