missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyclopropanecarboxylate 98.0+%, TCI America™

Catalog No. C106025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C106025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C106025ML Supplier TCI America Supplier No. C106025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4606-07-9
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00001282
InChI Key LDDOSDVZPSGLFZ-UHFFFAOYSA-N
Synonym cyclopropanecarboxylic acid ethyl ester, ethyl cyclopropylcarboxylate, ethyl cyclopropane carboxylate, cyclopropanecarboxylic acid, ethyl ester, pubchem11840, ethyl cylopropylcarboxylate, ethoxycarbonyl cyclopropane, ethylcyclopropane carboxylate, ksc497s5p, lddosdvzpsglfz-uhfffaoysa
PubChem CID 78351
IUPAC Name ethyl cyclopropanecarboxylate
SMILES CCOC(=O)C1CC1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 3272
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cyclopropanecarboxylate
EINECSNumber (3)-2440
TSCA No