missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Bromofluoroacetate 98.0+%, TCI America™

Catalog No. B16735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16735G 5 g
B167325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16735G Supplier TCI America Supplier No. B16735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 401-55-8
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.992
MDL Number MFCD00042095
InChI Key ULNDTPIRBQGESN-UHFFFAOYSA-N
Synonym ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t
PubChem CID 2733407
IUPAC Name ethyl 2-bromo-2-fluoroacetate
SMILES CCOC(=O)C(F)Br

Specifications

Color Yellow
Boiling Point 68°C
Quantity 5 g
UN Number 2810
Formula Weight 184.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Bromofluoroacetate