missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthyle benzoylacétatate 95,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 270°C |
| Quantity | 25 g |
| Formula Weight | 192.21 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | Ethyl Benzoylacetate |
Sécurité et manipulation
| EINECSNumber | (3)-1282&(3)-1708&(3)-1743 |
| RTECSNumber | AF4878000 |
| TSCA | Yes |