missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18602497
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602497 500 mL Amber Glass Bottle
18602498 6 x 500 mL Amber Glass Bottle
18602499 4 L Amber Glass Bottle
18602500 4 x 4 L Amber Glass Bottle
18602501 20 L Steel Can
18602502 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602497 Supplier Spectrum Chemical Mfg Cor Supplier No. ET105500MLGL
Only null left

ET105, 141-78-6, C4H8O2

Spectrum™ Chemical Ethyl Acetate, NF is used as a diluent and solvent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
UN Number UN1173
Boiling Point 77°C
Percent Purity 98 to 102%
Grade NF
Chemical Name or Material Ethyl Acetate