missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate HPLC, Lab Chem

Catalog No. LC141304
Change view
Click to view available options
Acidity:
<0.0009 meq/g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Acidity
LC141304 <0.0009 meq/g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC141304 Supplier LabChem Supplier No. LC141304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Ethyl Acetate HPLC Solvent, Safety: Toxic Flammable, Irritant

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
IUPAC Name Ethyl Acetate

Specifications

Color Colorless
Acidity <0.0009 meq/g
Appearance Colorless Liquid
Density 0.8946 g/cm3
Quantity 4 L
Ignition Point 426 °C
Odor Fruity
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation < 3 ppm
Specific Gravity 0.8946
Titratable Acid <0.0009 meq/g
Vapor Pressure 91.84 hPa at 18.7 C
Linear Formula CH3CO2CH2CH3
Water <0.05%
UN Number UN1173
Boiling Point 77 °C
Solubility Information Immisible in water
Formula Weight 88.1051
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material Ethyl Acetate HPLC
Show More Show Less
Hazard Category Flammable
Hazard Statement Eye Irritation
ShelfLife 5 Years
DOTInformation UN1173