missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, ≥99.5%, Solstice
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.8945 g/cm3 |
| Quantity | 4 x 4 L |
| Flash Point | −4°C |
| Packaging | Poly-coated Bottle |
| Water | 0.03% max. |
| UN Number | UN1173 |
| Beilstein | 506104 |
| Solubility Information | 870% in Water at 20°C |
| Boiling Point | 77°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate |
| EINECSNumber | 205-500-4 |