missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%

Catalog No. AAB2106614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2106614 25 g
AAB2106603 1 g
AAB2106606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2106614 Supplier Thermo Scientific Chemicals Supplier No. B2106614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50551-58-1 | C12H12O4 | 220.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50551-58-1
Molecular Formula C12H12O4
Molecular Weight (g/mol) 220.22
MDL Number MFCD00060510
InChI Key SXCMZYVXJUJIDJ-UHFFFAOYSA-N
PubChem CID 521284
IUPAC Name ethyl 7-methoxy-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC(OC)=C2O1

Specifications

Melting Point 83°C to 86°C
Quantity 25 g
Synonym ethyl 7-methoxybenzofuran-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acid ethyl ester, 7-methoxybenzofuran-2-carboxylic acid, ethyl ester, ethyl 7-methoxybenzo b furan-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acidethyl ester, ethyl7-methoxybenzofuran-2-carboxylate, acmc-1aowz, 2-ethoxycarbonyl-7-methoxybenzofuran, ethyl 4-methoxylbenzofunan-2-carboxylate, ethyl 7-methoxybenzo d furan-2-carboxylate
Formula Weight 220.22
Percent Purity 97%
Chemical Name or Material Ethyl 7-methoxybenzo[b]furan-2-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only