missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%

Catalog No. AAH25763MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25763MD 250 mg
AAH2576303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25763MD Supplier Thermo Scientific Chemicals Supplier No. H25763MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2037-81-2 | C12H11N3O4S | 293.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2037-81-2
Molecular Formula C12H11N3O4S
Molecular Weight (g/mol) 293.30
MDL Number MFCD00100230
InChI Key FCPHNPRVMWSGSK-UHFFFAOYSA-N
Synonym ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, ethyl 5-phenoxycarbonyl amino-1,2,3-thiadiazole-4-carboxylate, 5-n-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, tos-bb-0370, ethyl 5-phenoxycarbonylamino thiadiazole-4-carboxylate, ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
PubChem CID 6917126
IUPAC Name ethyl 5-(phenoxycarbonylamino)thiadiazole-4-carboxylate
SMILES CCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1

Specifications

Melting Point 153°C to 155°C
Quantity 250 mg
Formula Weight 293.3
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only