missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromovalerate, 98%

CAS: 14660-52-7 | C7H13BrO2 | 209.08 g/mol

Supplier:  Thermo Scientific Chemicals A1195222

 View more versions of this product

Catalog No. AAA1195222


Only null left
Explore available promotions

Description

Description

Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
Specifications

Specifications

Ethyl 5-bromovalerate
1.306
103°C (217°F)
1.458
MFCD00000266
1752023
ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
AFRWBGJRWRHQOV-UHFFFAOYSA-N
ethyl 5-bromopentanoate
84580
98%
14660-52-7
227°C
C7H13BrO2
Br(CH2)4CO2CH2CH3
100 g
Light sensitive
Not miscible or difficult to mix in water.
CCOC(=O)CCCCBr
209.08
209.09

Chemical Identifiers

14660-52-7
209.08
AFRWBGJRWRHQOV-UHFFFAOYSA-N
ethyl 5-bromopentanoate
C7H13BrO2
MFCD00000266
84580
CCOC(=O)CCCCBr
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-705-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only