missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromovalerate, 98%

Catalog No. p-7081804
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1195214 25 g
AAA1195222 100 g
AAA1195236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1195214 Supplier Thermo Scientific Chemicals Supplier No. A1195214

CAS: 14660-52-7 | C7H13BrO2 | 209.08 g/mol

Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.

Chemical Identifiers

CAS 14660-52-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00000266
InChI Key AFRWBGJRWRHQOV-UHFFFAOYSA-N
PubChem CID 84580
IUPAC Name ethyl 5-bromopentanoate
SMILES CCOC(=O)CCCCBr

Specifications

Density 1.306
Boiling Point 227°C
Flash Point 103°C (217°F)
Refractive Index 1.458
Linear Formula Br(CH2)4CO2CH2CH3
Quantity 25 g
Beilstein 1752023
Sensitivity Light sensitive
Synonym ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 209.09
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromovalerate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-705-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only