missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Bromothiophene-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7129337
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11111G 1 g
E11115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11111G Supplier TCI America Supplier No. E11111G

Chemical Identifiers

CAS 5751-83-7
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.095
MDL Number MFCD02683089
InChI Key PZNHMXAOMDQLLE-UHFFFAOYSA-N
Synonym 5-bromothiophene-2-carboxylic acid ethyl ester, ethyl 5-bromo-2-thiophenecarboxylate, ethyl5-bromothiophene-2-carboxylate, 5-bromo-thiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-bromo-, ethyl ester, pubchem16118, acmc-209zz5, ksc494s9b, 2-bromo-5-ethoxycarbonylthiophene, ethyl 5-bromothiophene-2carboxylate
PubChem CID 605723
IUPAC Name ethyl 5-bromothiophene-2-carboxylate
SMILES CCOC(=O)C1=CC=C(S1)Br

Specifications

Color Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 235.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 5-Bromothiophene-2-carboxylate