missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Oxazolecarboxylate 96.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 48°C |
| Color | White-Yellow |
| Boiling Point | 101°C |
| Quantity | 1 g |
| Formula Weight | 141.13 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethyl 4-Oxazolecarboxylate |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |