missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Methoxyphenylacetate 98.0+%, TCI America™

Numéro de catalogue. M079325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M079325ML 25 mL
M0793500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M079325ML Fournisseur TCI America Code fournisseur M079325ML
Il en reste null

Identifiants chimiques

CAS 14062-18-1
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00040760
InChI Key DOCCDOCIYYDLGJ-UHFFFAOYSA-N
Synonym ethyl 2-4-methoxyphenyl acetate, ethyl 4-methoxyphenylacetate, 4-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-methoxy-, ethyl ester, ethyl p-methoxyphenylacetate, ethyl 4-methoxyphenyl acetate, 4-methoxy-phenyl-acetic acid ethyl ester, acmc-1bnx2, homoanisic acid ethyl ester
PubChem CID 84174
IUPAC Name ethyl 2-(4-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)OC

Spécifications

Color Yellow
Boiling Point 158°C
Quantity 25 mL
UN Number 1993
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Methoxyphenylacetate