missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. E08845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08845G 5 g
E088425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08845G Supplier TCI America Supplier No. E08845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002353
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
Synonym ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
PubChem CID 8434
ChEBI CHEBI:86616
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 117°C
Color White
Boiling Point 178°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Hydroxybenzoate [for Biochemical Research]
EINECSNumber (3)-1585
RTECSNumber DH2190000
TSCA Yes