missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxybenzoate, 99%

Catalog No. AAA1317230
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1317230 250 g
AAA131720B 1000 g
AAA131720I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1317230 Supplier Thermo Scientific Chemicals Supplier No. A1317230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120-47-8 | C9H10O3 | 166.176 g/mol

Ethyl 4-hydroxybenzoate (ethylparaben) was used to study the in-situderivatisation solid-phase microextraction procedure for determining parabens, related chlorophenols and triclosan in water. It is a novel sorbent for solid-phase extraction, was used to study its retention property. It has high extraction efficiency towards the compounds tested owing to the electrostatic interaction, hydrophobic interaction and hydrogen bonding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-hydroxybenzoate (ethylparaben) was used to study the in-situderivatisation solid-phase microextraction procedure for determining parabens, related chlorophenols and triclosan in water. It is a novel sorbent for solid-phase extraction, was used to study its retention property. It has high extraction efficiency towards the compounds tested owing to the electrostatic interaction, hydrophobic interaction and hydrogen bonding.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002353
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
Synonym ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
PubChem CID 8434
ChEBI CHEBI:86616
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 116°C to 118°C
Boiling Point 297°C to 298°C (decomposition)
Quantity 250 g
Beilstein 1101972
Merck Index 14,3837
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl 4-hydroxybenzoate
EINECSNumber 204-399-4
RTECSNumber DH2190000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only