missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (4-bromobenzoyl)acetate, 95%, Thermo Scientific™

Numéro de catalogue. p-4089899
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC399160010 1g Glass bottle
AC399160050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC399160010 Fournisseur Thermo Scientific Chemicals Code fournisseur 399160010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 26510-95-2
Formule moléculaire C11H11BrO3
Poids moléculaire (g/mol) 271.1
Numéro MDL MFCD00231243
Clé InChI PBDYXCKRDRCJDC-UHFFFAOYSA-N
Synonyme ethyl 3-4-bromophenyl-3-oxopropanoate, ethyl 4-bromobenzoyl acetate, ethyl 4-bromobenzoylacetate, 3-4-bromo-phenyl-3-oxo-propionic acid ethyl ester, 3-4-bromophenyl-3-oxo-propionic acid ethyl ester, ethyl 3-4-bromophenyl-3-oxo-propionate, benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester, ethyl p-bromobenzoyl acetate, benzenepropanoic acid,4-bromo-b-oxo-, ethyl ester
PubChem CID 2757149
Nom de l’IUPAC ethyl 3-(4-bromophenyl)-3-oxopropanoate
SOURIRES CCOC(=O)CC(=O)C1=CC=C(C=C1)Br

Spécifications

Nom ou substance chimique Ethyl (4-bromobenzoyl)acetate
Densité 1.4320g/mL
Point d’ébullition 268°C to 269°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 94% min. (HPLC)
Emballage Glass bottle
Formule linéaire BrC6H4C(O)CH2CO2CH2CH3
Quantité 1g
Beilstein 10, I, 324
Gravité spécifique 1.432
Poids de formule 271.1
Pourcentage de pureté 95%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only