missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate, 97%, Thermo Scientific Chemicals

Catalog No. AAL1996906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1996906 Supplier Thermo Scientific Chemicals Supplier No. L1996906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91600-33-8 | Molecular Formula: C7H11F3O3 | Molecular Weight: 200.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91600-33-8
Molecular Formula C7H11F3O3
Molecular Weight (g/mol) 200.157
MDL Number MFCD00190634
InChI Key MFRIOKNLYRUYHP-UHFFFAOYSA-N
Synonym ethyl 2-methyl-3-hydroxy-4,4,4-trifluorobutyrate, ethyl 3-hydroxy-2-methyl-4,4,4-trifluorobutyrate, ethyl 2-methyl-4,4,4-trifluoro-3-hydroxybutanoate, ethyl-4,4,4-trifluor-3-hydroxy-2-methylbutanoat, ethyl 2-methyl-4,4,4-trifluoro-3-hydroxybutyrate, ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate, ethyl 3-hydroxy-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate, 4,4,4-trifluoro-3-hydroxy-2-methylbutyric acid ethyl ester
PubChem CID 533787
IUPAC Name ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutanoate
SMILES CCOC(=O)C(C)C(C(F)(F)F)O

Specifications

Boiling Point 180°C to 184°C
Quantity 5 g
Beilstein 6194166
Formula Weight 200.16
Percent Purity 97%
Chemical Name or Material Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only