missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(Trifluoromethyl)pyrazole-4-carboxylate 98.0+%, TCI America™

Catalog No. E10245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10245G 5 g
E10241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10245G Supplier TCI America Supplier No. E10245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 155377-19-8
Molecular Formula C7H7F3N2O2
Molecular Weight (g/mol) 208.14
MDL Number MFCD00052083
InChI Key VYXIHSAEOXPAEY-UHFFFAOYSA-N
Synonym ethyl 3-trifluoromethyl pyrazole-4-carboxylate, ethyl 3-trifluoromethyl-1h-pyrazole-4-carboxylate, ethyl 5-trifluoromethyl-1h-pyrazole-4-carboxylate, 3-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-trifluoromethyl pyrazole-4-carboxylic acid ethyl ester, ethyl 3-trifluoromethyl-2h-pyrazole-4-carboxylate, 5-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 1h-pyrazole-4-carboxylic acid, 3-trifluoromethyl-, ethyl ester
PubChem CID 596095
IUPAC Name ethyl 5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)C(F)(F)F

Specifications

Melting Point 145°C
Color Yellow
Quantity 5 g
Formula Weight 208.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-(Trifluoromethyl)pyrazole-4-carboxylate