missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Phenylglycidate (cis- and trans- mixture) 90.0+%, TCI America™

Catalog No. E079825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E079825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E079825G Supplier TCI America Supplier No. E079825G

Chemical Identifiers

CAS 121-39-1
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00005123
InChI Key GOMAKLPNAAZVCJ-UHFFFAOYSA-N
Synonym ethyl 3-phenylglycidate, ethyl phenylglycidate, oxiranecarboxylic acid, 3-phenyl-, ethyl ester, ethyl 2,3-epoxy-3-phenylpropionate, ethyl 3-phenyloxiranecarboxylate, glycidic acid, 3-phenyl-, ethyl ester, 2-ethyl-3-phenylglycidate, fema no. 2454, ccris 2623, ethyl3-phenylglycidate
PubChem CID 8469
IUPAC Name ethyl 3-phenyloxirane-2-carboxylate
SMILES CCOC(=O)C1C(O1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 192.21
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Phenylglycidate (cis- and trans- mixture)
EINECSNumber (3)-1729
RTECSNumber MB4970000
TSCA Yes
Recommended Storage Refrigerator