missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Oxo-3-(4-chlorophenyl)propionate 98.0+%, TCI America™

Catalog No. p-7129310
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10591G 1 g
E10595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10591G Supplier TCI America Supplier No. E10591G

Chemical Identifiers

CAS 2881-63-2
Molecular Formula C11H11ClO3
Molecular Weight (g/mol) 226.656
MDL Number MFCD00018713
InChI Key DGCZHKABHPDNCC-UHFFFAOYSA-N
Synonym 3-Oxo-3-(4-chlorophenyl)propionic Acid Ethyl Ester
PubChem CID 101336
IUPAC Name ethyl 3-(4-chlorophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 123°C
Quantity 1 g
Formula Weight 226.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 3-Oxo-3-(4-chlorophenyl)propionate
TSCA No
Recommended Storage Refrigerator