missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-nitrobenzoate, 98+%

Catalog No. AAA1062622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062622 100 g
AAA1062614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1062622 Supplier Thermo Scientific Chemicals Supplier No. A1062622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-98-4 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-98-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00014702
InChI Key MKBIJCPQTPFQKQ-UHFFFAOYSA-N
Synonym ethyl m-nitrobenzoate, benzoic acid, 3-nitro-, ethyl ester, ethyl-m-nitrobenzoate, m-nitrobenzoic acid, ethyl ester, benzoic acid, m-nitro-, ethyl ester, acmc-20akzc, 3-nitrobenzoic acid ethyl, benzoic acid, 3-nitro-,ethyl ester, benzoic acid, m-nitro-, ethyl ester 8ci, r---2-2-diphenylphosphino phenyl-4-phenyl-2-oxazoline
PubChem CID 69261
IUPAC Name ethyl 3-nitrobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 40°C to 43°C
Boiling Point 297°C to 298°C
Quantity 100 g
Beilstein 1912880
Merck Index 14,3832
Formula Weight 195.18
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-nitrobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-574-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only