missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Methylpyrazole-5-carboxylate 98.0+%, TCI America™

Catalog No. E11005G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11005G 5 g
E11001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11005G Supplier TCI America Supplier No. E11005G

Chemical Identifiers

CAS 4027-57-0
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00052514
InChI Key BOTXQJAHRCGJEG-UHFFFAOYSA-N
PubChem CID 77645
IUPAC Name ethyl 5-methyl-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NNC(C)=C1

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 5 g
Synonym ethyl 3-methyl-1h-pyrazole-5-carboxylate, ethyl 3-methylpyrazole-5-carboxylate, 5-methyl-1h-pyrazole-3-carboxylic acid ethyl ester, 3-methyl-1h-pyrazole-5-carboxylic acid ethyl ester, 1h-pyrazole-3-carboxylic acid, 5-methyl-, ethyl ester, ethyl3-methylpyrazole-5-carboxylate, 5-methyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl3-methyl-1h-pyrazole-5-carboxylate, ethyl 5-methyl-2h-pyrazole-3-carboxylate, pubchem7843
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Methylpyrazole-5-carboxylate