missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthyle 3-Mercaptopropionate 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Quantity | 25 g |
| Formula Weight | 134.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Ethyl 3-Mercaptopropionate |
Sécurité et manipulation
| TSCA | Yes |