missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isothiocyanatopropionate, 97%

Catalog No. AAL1091106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1091106 5 g
AAL1091103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1091106 Supplier Thermo Scientific Chemicals Supplier No. L1091106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17126-62-4 | C6H9NO2S | 159.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17126-62-4
Molecular Formula C6H9NO2S
Molecular Weight (g/mol) 159.203
MDL Number MFCD00041130
InChI Key UPTRONYNXNYITM-UHFFFAOYSA-N
Synonym ethyl 3-isothiocyanatopropionate, 3-isothiocyanato-propionic acid ethyl ester, acmc-1ca2j, ethyl-3-isothiocynato propionate, isothiocyanato-propionic acid ethyl ester, 3-isothiocyanatopropionic acid ethyl ester, propanoic acid,3-isothiocyanato-,ethyl ester, propanoic acid,3-isothiocyanato-, ethyl ester
PubChem CID 526514
IUPAC Name ethyl 3-isothiocyanatopropanoate
SMILES CCOC(=O)CCN=C=S

Specifications

Density 1.12
Boiling Point 75°C to 76°C (8 mmHg)
Odor Pungent
Refractive Index 1.5014
Linear Formula SCNCH2CH2CO2CH2CH3
Quantity 5 g
UN Number UN2922
Beilstein 1705633
Sensitivity Moisture sensitive
Formula Weight 159.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-isothiocyanatopropionate
Show More Show Less
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only